Sodium hexafluoroantimonate ≥98%

Supplier: BeanTown Chemical

Synonyms: Sodium hexafluoroantimonate(V)

142510-100G 142510-25G 142510-500G
BT142510-100GEA 107 USD
BT142510-100G BT142510-25G BT142510-500G
Sodium hexafluoroantimonate ≥98%
Sodium hexafluoroantimonate

CAS: 16925-25-0; EC No: 240-989-8; MDL No: MFCD00003482 UN No: UN1549; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: NaSbF6; MW: 258.73 Density (g/mL): 3.375 Air Sensitive, Moisture Sensitive


  • Air Sensitive, Moisture Sensitive
  • Powder

Formula: F₆NaSb
Density: 3.375 g/mL
MDL Number: MFCD00003482
CAS Number: 16925-25-0
EINECS: 240-989-8
UN: 1549
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR