1,2-Difluoro-4-nitrobenzene ≥98%
Supplier: BeanTown Chemical
Synonyms:
3,4-Difluoronitrobenzene
142500-10G
142500-50G
BT142500-10GEA
38
USD
BT142500-10G
BT142500-50G
1,2-Difluoro-4-nitrobenzene ≥98%
1,2-Difluoro-4-nitrobenzene
CAS: 369-34-6; EC No: 206-718-2; MDL No: MFCD00007198; RTECS: CZ5710000
UN No: UN2810; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: C6H5FN2O2; MW: 159.09
Boiling Point: 76-80°/11 mmHg; Flash point: 81°C (178°F)
Density (g/mL): 1.437; Refractive Index: 1.509
- RTECS: CZ5710000
- Refractive index: 1.509
- Liquid
Formula:
C₆H₃F₂NO₂ Boiling Pt: 76…80 °C/11 mmHg Density: 1.437 g/mL Flash Pt: 81 °C (178 °F) |
MDL Number:
MFCD00007198 CAS Number: 369-34-6 EINECS: 206-718-2 UN: 2810 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...