Copper(I) chloride ≥97%

Supplier: BeanTown Chemical

Synonyms: Cuprous chloride, Copper chloride

142360-100G 142360-2KG 142360-500G
BT142360-100GEA 31.45 USD
BT142360-100G BT142360-2KG BT142360-500G
Copper(I) chloride ≥97%
Copper (I) chloride

CAS: 7758-89-6; EC No: 231-842-9; MDL No: MFCD00010971; RTECS: GL6990000 UN No: UN2802; Haz Class: 8; Packing Group: III Powder; Linear Formula: CuCl; MW: 98.99 Melting Point: 430°; Boiling Point: 1490° Density (g/mL): 4.14 Air Sensitive, Moisture Sensitive


  • RTECS: GL6990000
  • Air Sensitive, Moisture Sensitive
  • Powder

Formula: ClCu
MW: 99 g/mol
Boiling Pt: 1367
Melting Pt: 430
Density: 4.14 g/mL
MDL Number: MFCD00010971
CAS Number: 7758-89-6
EINECS: 231-842-9
UN: 2802
ADR: 8 III
Merck Index: 13,02690

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR