Telluric acid dihydrate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Hydrogen tellurate
141460-10G
141460-50G
BT141460-10GEA
49.81
USD
BT141460-10G
BT141460-50G
Telluric acid dihydrate ≥99%
Telluric acid dihydrate
CAS: 13451-14-4; EC No: 232-267-6; MDL No: MFCD00150387
UN No: UN3284; Haz Class: 6.1; Packing Group: III
Crystalline; Linear Formula: Te(OH)6; MW: 229.64
Melting Point: 136°; Boiling Point: 160° (decomposes)
Density (g/mL): 3.15
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
H₂O₄Te.2H₂0 MW: 229.64 g/mol (193.61 anhy) Boiling Pt: 160 °C (dec.) Melting Pt: 136 °C Density: 3.15 g/mL |
MDL Number:
MFCD00150387 CAS Number: 13451-14-4 EINECS: 232-267-6 ADR: 6.1 III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...