Methylcyclopentadienylmanganese tricarbonyl ≥97%

Supplier: BeanTown Chemical

141065-1G 141065-25G 141065-5G
BT141065-1GEA 25 USD
BT141065-1G BT141065-25G BT141065-5G
Methylcyclopentadienylmanganese tricarbonyl ≥97%
Methylcyclopentadienylmanganese tricarbonyl

CAS: 12108-13-3; EC No: 235-166-5; MDL No: MFCD00001426 UN No: UN3281; Haz Class: 6.1; Packing Group: II Liquid; Linear Formula: C5H4CH3Mn(CO)3; Molecular Formula: C9H7MnO3; MW: 218.09 Melting Point: -1°; Boiling Point: 232-233°; Flash point: 96°C (205°F) Density (g/mL): 1.388; Refractive Index: 1.584 Air Sensitive


  • Refractive index: 1.584
  • Air Sensitive
  • Liquid

Formula: C₉H₇MnO₃
Boiling Pt: 232…233 °C
Melting Pt: –1 °C
Density: 1.388 g/mL
Flash Pt: 96 °C (205 °F)
MDL Number: MFCD00001426
CAS Number: 12108-13-3
EINECS: 235-166-5
UN: 3281
ADR: 6.1,II

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR