Pyridinium tribromide ≥90%, Technical Grade

Supplier: BeanTown Chemical

Synonyms: Pyridinium bromide perbromide, Monopyridin-1-ium tribromide, Pyridine hydrobromide perbromide

140975-100G 140975-25G 140975-500G
BT140975-100GEA 94.79 USD
BT140975-100G BT140975-25G BT140975-500G
Pyridinium tribromide ≥90%, Technical Grade
Pyridinium tribromide

CAS: 39416-48-3; EC No: 254-446-8; MDL No: MFCD00013223 UN No: UN3261; Haz Class: 8; Packing Group: III Crystalline; Molecular Formula: C5H5N·HBr·Br2; MW: 319.82 Melting Point: 128-136°


  • Crystalline

Formula: C₅H₆Br₃N
MW: 319.82 g/mol
Melting Pt: 128…136 °C
MDL Number: MFCD00013223
CAS Number: 39416-48-3
EINECS: 254-446-8
UN: 3261
ADR: 8 III

Order Now

Specification Test Results

Assay 90%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR