Rubidium hydroxide hydrate ≥99%

Supplier: BeanTown Chemical

140910-25G 140910-5G
BT140910-25GEA 293 USD
BT140910-25G BT140910-5G
Rubidium hydroxide hydrate ≥99%
Rubidium hydroxide hydrate

CAS: 12026-05-0; EC No: 215-186-0; MDL No: MFCD00011194 UN No: UN2678; Haz Class: 8; Packing Group: II Crystalline/Lump; Linear Formula: RbOH·xH2O; MW: 102.48 (anhydrous) Air Sensitive


  • Air Sensitive
  • Crystalline/Lump

Formula: H₃O₂Rb
MDL Number: MFCD00149835
CAS Number: 12026-05-0
EINECS: 215-186-0
ADR: 8,II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR