Sodium orthosilicate

Supplier: BeanTown Chemical
Danger

Synonyms: Tetrasodium orthosilicate, Sodium silicate (ortho)

140900-1KG 140900-5KG
BT140900-1KGEA 44 USD
BT140900-1KG BT140900-5KG
Sodium orthosilicate
tetra-Sodium orthosilicate

CAS: 13472-30-5; MDL No: MFCD00014252 UN No: UN1759; Haz Class: 8; Packing Group: II Crystalline; Linear Formula: Na4SiO4; MW: 184.04


  • Crystalline

Formula: Na₄O₄Si
MW: 184.04 g/mol
Melting Pt: 1078 °C
Storage Temperature: Ambient
MDL Number: MFCD00014252
CAS Number: 13472-30-5
UN: 1759
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR