Oxalic acid, anhydrous ≥98%

Supplier: BeanTown Chemical

Synonyms: Ethanedioic acid

140220-250G 140220-50G
BT140220-250GEA 58.77 USD
BT140220-250G BT140220-50G
Oxalic acid, anhydrous ≥98%
Oxalic acid

CAS: 144-62-7; EC No: 205-634-3; MDL No: MFCD00002573; RTECS: RO2450000 UN No: UN3261; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: HOOCCOOH; Molecular Formula: C2H2O4; MW: 90.04 Melting Point: 190° (decomposes); Boiling Point: 157° (sublimes) Density (g/mL): 1.9


  • RTECS: RO2450000
  • Crystalline

Formula: C₂H₂O₄
MW: 90.04 g/mol
Boiling Pt: 365 (subl.)
Melting Pt: 190 °C (dec.)
Density: 1.9 g/mL
MDL Number: MFCD00002573
CAS Number: 144-62-7
EINECS: 205-634-3
UN: 3261
ADR: 8 III
Merck Index: 13,06980

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR