Benzyl azide ≥94%

Supplier: BeanTown Chemical

139200-1G 139200-25G 139200-5G
BT139200-1GEA 28.64 USD
BT139200-1G BT139200-25G BT139200-5G
Benzyl azide ≥94%
Benzyl azide

CAS: 622-79-7; MDL No: MFCD00013836 UN No: UN1993; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C6H5CH2N3; MW: 133.15 Boiling Point: 82-85°/16 mmHg; Flash point: 43°C (110°F) Density (g/mL): 1.066


  • Liquid

Formula: C₇H₇N₃
Boiling Pt: 82…85 °C/16 mmHg
Density: 1.066 g/mL
Flash Pt: 43 °C (110 °F)
MDL Number: MFCD00013836
CAS Number: 622-79-7
ADR: 3,III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR