Hexafluorobenzene ≥99%
Supplier: BeanTown Chemical
Synonyms:
Perfluorobenzene
138905-10G
138905-250G
138905-50G
BT138905-10GEA
51.15
USD
BT138905-10G
BT138905-250G
BT138905-50G
Hexafluorobenzene ≥99%
Hexafluorobenzene
CAS: 392-56-3; EC No: 206-876-2; MDL No: MFCD00000288; RTECS: DA3050000
UN No: UN1993; Haz Class: 3; Packing Group: II
Liquid; Molecular Formula: C6F6; MW: 186.06
Melting Point: 4-5°; Boiling Point: 80-82°; Flash point: 10°C (50°F)
Density (g/mL): 1.612; Refractive Index: 1.377
- RTECS: DA3050000
- Refractive index: 1.377
- Liquid
Formula:
C₆F₆ MW: 186.06 g/mol Boiling Pt: 80…82 °C Melting Pt: 4…5 °C Density: 1.612 g/mL Flash Pt: 10 °C (50 °F) |
MDL Number:
MFCD00000288 CAS Number: 392-56-3 EINECS: 206-876-2 UN: 1993 ADR: 3,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...