Sodium dodecyl sulfate (SDS) ≥99% (dry weight)

Supplier: BeanTown Chemical

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

138190-100G 138190-25G 138190-500G
BT138190-100GEA 78.21 USD
BT138190-100G BT138190-25G BT138190-500G
Sodium dodecyl sulfate (SDS) ≥99% (dry weight)
Sodium dodecyl sulphate (SDS)

CAS: 151-21-3; EC No: 205-788-1; MDL No: MFCD00036175; RTECS: WT1050000 UN No: UN2926; Haz Class: 4.1 (6.1); Packing Group: II Solid; Linear Formula: CH3(CH2)11OSO3Na; Molecular Formula: C12H25NaO4S; MW: 288.38 Melting Point: 204-207°; Flash point: 170°C (338°F) Density (g/mL): 0.37


  • RTECS: WT1050000
  • Solid

Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Melting Pt: 204…207 °C
Density: 0.37 g/mL
Flash Pt: 170 °C (338 °F)
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 2926
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR