1,4,7,10-Tetraazacyclododecane tetrahydrochloride ≥98%

Supplier: BeanTown Chemical

Synonyms: Cyclen tetrahydrochloride

137700-1G 137700-250MG 137700-5G
BT137700-1GEA 217 USD
BT137700-1G BT137700-250MG BT137700-5G
1,4,7,10-Tetraazacyclododecane tetrahydrochloride ≥98%
1,4,7,10-Tetraazacyclododecane tetrahydrochloride

CAS: 10045-25-7; MDL No: MFCD00068484 UN No: UN3077; Haz Class: 9; Packing Group: III Solid; Molecular Formula: C8H20N4·4HCl; MW: 318.11 Melting Point: 293° (decomposes) Hygroscopic


  • Hygroscopic
  • Solid

Formula: C₈H₂₄Cl₄N₄
Melting Pt: 293 °C (dec.)
MDL Number: MFCD00068484
CAS Number: 10045-25-7
ADR: 9,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR