Potassium hexafluorophosphate ≥98%
Supplier: BeanTown Chemical
137435-1KG
137435-250G
137435-50G
BT137435-1KGEA
302.74
USD
BT137435-1KG
BT137435-250G
BT137435-50G
Potassium hexafluorophosphate ≥98%
Potassium hexafluorophosphate
CAS: 17084-13-8; EC No: 241-143-0; MDL No: MFCD00011412
UN No: UN3260; Haz Class: 8; Packing Group: II
Crystalline; Linear Formula: KPF6; MW: 184.07
Melting Point: 575°
Density (g/mL): 2.75
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
F₆KP MW: 184.06 g/mol Melting Pt: 575 °C Density: 2.75 g/mL |
MDL Number:
MFCD00011412 CAS Number: 17084-13-8 EINECS: 241-143-0 UN: 3260 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...