Silver perchlorate monohydrate ≥99.9% (trace metals basis)
Supplier: BeanTown Chemical
137220-25G
137220-5G
BT137220-25GEA
206
USD
BT137220-25G
BT137220-5G
Silver perchlorate monohydrate ≥99.9% (trace metals basis)
Silver perchlorate monohydrate
CAS: 14242-05-8; EC No: 232-035-4; MDL No: MFCD00149128
UN No: UN1481; Haz Class: 5.1; Packing Group: II
Crystalline; Molecular Formula: AgClO4·H2O; MW: 225.34
Melting Point: 485° (decomposes)
Density (g/mL): 2.8
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
AgClO₄·1H₂O Melting Pt: 485 °C (dec.) Density: 2.8 g/mL |
MDL Number:
MFCD00149128 CAS Number: 14242-05-8 EINECS: 232-035-4 UN: 1481 ADR: 5.1,II |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...