Platinum(II) chloride ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Platinum dichloride

136605-1G 136605-5G
BT136605-1GEA 253.53 USD
BT136605-1G BT136605-5G
Platinum(II) chloride ≥99.9%
Platinum (II) chloride

CAS: 10025-65-7; EC No: 233-034-1; MDL No: MFCD00011181; RTECS: TP2275000 UN No: UN3260; Haz Class: 8; Packing Group: III Crystalline/Powder; Linear Formula: PtCl2; MW: 265.99 Melting Point: 581° (decomposes) Density (g/mL): 5.87


  • RTECS: TP2275000
  • Crystalline/Powder

Formula: Cl₂Pt
MW: 265.99 g/mol
Melting Pt: 581 (dec.)
Density: 5.87 g/mL
MDL Number: MFCD00011181
CAS Number: 10025-65-7
EINECS: 233-034-1
UN: 3260
ADR: 8 III
Merck Index: 12,07687

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR