Triphosgene ≥98%

Supplier: BeanTown Chemical

Synonyms: Bis(trichloromethyl) carbonate

135635-100G 135635-25G 135635-5G
BT135635-100GEA 447 USD
BT135635-100G BT135635-25G BT135635-5G
Triphosgene ≥98%
Triphosgene

CAS: 32315-10-9; EC No: 250-986-3; MDL No: MFCD00062848 UN No: UN2928; Haz Class: 6.1 (8); Packing Group: II Solid; Linear Formula: Cl3COCOOCCl3; Molecular Formula: C3Cl6O3; MW: 296.75 Melting Point: 79-83°; Boiling Point: 203-206° Density (g/mL): 1.6 Heat Sensitive, Moisture Sensitive


  • Heat Sensitive, Moisture Sensitive
  • Solid

Formula: C₃Cl₆O₃
MW: 296.75 g/mol
Boiling Pt: 203…206 °C
Melting Pt: 79…83 °C
Density: 1.6 g/mL
MDL Number: MFCD00062848
CAS Number: 32315-10-9
EINECS: 250-986-3
UN: 2928
ADR: 6.1,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR