Nitromethane ≥98%

Supplier: BeanTown Chemical

135205-100G 135205-2.5KG 135205-500G
BT135205-100GEA 36.19 USD
BT135205-100G BT135205-2.5KG BT135205-500G
Nitromethane ≥98%
Nitromethane

CAS: 75-52-5; EC No: 200-876-6; MDL No: MFCD00007400; RTECS: PA9800000 UN No: UN1261; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: CH3NO2; MW: 61.04 Melting Point: -29°; Boiling Point: 101°; Flash point: 35°C (95°F) Density (g/mL): 1.127; Refractive Index: 1.382


  • RTECS: PA9800000
  • Refractive index: 1.382
  • Liquid

Formula: CH₃NO₂
MW: 61.04 g/mol
Boiling Pt: 101 °C
Melting Pt: –29 °C
Density: 1.127 g/mL
Flash Pt: 35 °C (95 °F)
MDL Number: MFCD00007400
CAS Number: 75-52-5
EINECS: 200-876-6
UN: 1261
ADR: 3,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR