Ethoxyacetylene 50% (w/w) in hexanes
Supplier: BeanTown Chemical
CAS: 927-80-0; EC No: 213-164-5; MDL No: MFCD00009247; RTECS: KN9900000
UN No: UN1993; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3CH2OC≡CH; Molecular Formula: C4H6O; MW: 70.09
Flash point: -28°C (-18°F)
Density (g/mL): 0.718
- RTECS: KN9900000
- Liquid
Formula:
C₄H₆O Density: 0.718 g/mL Flash Pt: –28 °C (–18 °F) |
MDL Number:
MFCD00009247 CAS Number: 927-80-0 EINECS: 213-164-5 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...