Bismuth ≥99.5% (trace metals basis), powder -325 mesh

Supplier: BeanTown Chemical

134100-100G 134100-500G
BT134100-100GEA 48 USD
BT134100-100G BT134100-500G
Bismuth ≥99.5% (trace metals basis), powder -325 mesh
Bismuth

CAS: 7440-69-9; EC No: 231-177-4; MDL No: MFCD00134033; RTECS: EB2600000 UN No: UN3089; Haz Class: 4.1; Packing Group: III Powder, apparent density 3.2-4.0g/cm3; Molecular Formula: Bi; MW: 208.98 Melting Point: 271°; Boiling Point: 1560° Density (g/mL): 9.8


  • RTECS: EB2600000
  • Powder, apparent density 3.2-4.0g/cm3

Formula: Bi
Boiling Pt: 1560 °C
Melting Pt: 271 °C
Density: 9.8 g/mL
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
UN: 3089
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR