Tetraammineplatinum(II) nitrate (3 - 4% Pt) (w/w) in water

Supplier: BeanTown Chemical

133580-C2G 133580-C500MG
BT133580-C2GEA 695 USD
BT133580-C2G BT133580-C500
Tetraammineplatinum(II) nitrate (3 - 4% Pt) (w/w) in water
Tetraammineplatinum(II) nitrate

CAS: 20634-12-2; EC No: 243-929-9; MDL No: MFCD00011624 UN No: UN3218; Haz Class: 5.1; Packing Group: III Liquid; Linear Formula: Pt(NH3)4(NO3)2; Molecular Formula: H12N6O6Pt; MW: 387.21 Density (g/mL): 1.05


  • Liquid

Formula: H₁₂N₆O₆Pt
Density: 1.05 g/mL
MDL Number: MFCD00011624
CAS Number: 20634-12-2
EINECS: 243-929-9
ADR: 5.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR