Sodium nitrate ≥98%

Supplier: BeanTown Chemical

132835-10KG 132835-2.5KG 132835-500G
BT132835-10KGEA 262.3 USD
BT132835-10KG BT132835-2.5KG BT132835-500G
Sodium nitrate ≥98%
Sodium nitrate

CAS: 7631-99-4; EC No: 231-554-3; MDL No: MFCD00011119; RTECS: WC5600000 UN No: UN1498; Haz Class: 5.1; Packing Group: III Solid; Linear Formula: NaNO3; MW: 84.99 Melting Point: 306° (decomposes); Boiling Point: 380° Density (g/mL): 2.261


  • RTECS: WC5600000
  • Solid

Formula: NaNO₃
MW: 84.99 g/mol
Boiling Pt: 380 °C
Melting Pt: 306 °C (dec.)
Density: 2.261 g/mL
MDL Number: MFCD00011119
CAS Number: 7631-99-4
EINECS: 231-554-3
UN: 1498
ADR: 5.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR