Zinc ≥97.5% (trace metals basis), powder APS 6-9 µm
Supplier: BeanTown Chemical
131695-1KG
131695-250G
BT131695-1KGEA
75.99
USD
BT131695-1KG
BT131695-250G
Zinc ≥97.5% (trace metals basis), powder APS 6-9 µm
Zinc
CAS: 7440-66-6; EC No: 231-175-3; MDL No: MFCD00011291; RTECS: ZG8600000
UN No: UN1436; Haz Class: 4.3 (4.2); Packing Group: II
Powder; Molecular Formula: Zn; MW: 65.39
Melting Point: 420°; Boiling Point: 907°
Density (g/mL): 7.133
- RTECS: ZG8600000
- Powder
Formula:
Zn MW: 65.39 g/mol Boiling Pt: 908 Melting Pt: 420 Density: 7.133 g/mL |
MDL Number:
MFCD00011291 CAS Number: 7440-66-6 EINECS: 231-175-3 UN: 1436 ADR: 4.3 II Merck Index: 12,10255 |
Specification Test Results
Assay | 97.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...