Germanium(II) iodide ≥99.99%

Supplier: BeanTown Chemical

Synonyms: Germanium diiodide

131605-1G 131605-250MG 131605-5G
BT131605-1GEA 202 USD
BT131605-1G BT131605-250MG BT131605-5G
Germanium(II) iodide ≥99.99%
Germanium (II) iodide

CAS: 13573-08-5; EC No: 236-998-1; MDL No: MFCD00135537 UN No: UN3260; Haz Class: 8; Packing Group: II -10 Mesh Powder; Molecular Formula: GeI2; MW: 326.45 Density (g/mL): 5.37 Air Sensitive, Moisture Sensitive


  • Air Sensitive, Moisture Sensitive
  • -10 Mesh Powder

Formula: GeI₂
Density: 5.37 g/mL
MDL Number: MFCD00135537
CAS Number: 13573-08-5
EINECS: 236-998-1
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR