Sodium bromate ≥99.5% (trace metals basis)
Supplier: BeanTown Chemical
CAS: 7789-38-0; EC No: 232-160-4; MDL No: MFCD00003476
UN No: UN1494; Haz Class: 5.1; Packing Group: II
Crystalline; Linear Formula: NaBrO3; MW: 150.9
Melting Point: 381° (decomposes)
Density (g/mL): 3.339; Refractive Index: 1.594
- Refractive index: 1.594
- Crystalline
Formula:
BrNaO₃ Melting Pt: 381 °C (dec.) Density: 3.339 g/mL |
MDL Number:
MFCD00003476 CAS Number: 7789-38-0 EINECS: 232-160-4 UN: 1494 ADR: 5.1,II |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...