Copper ≥99.999% (trace metals basis), powder -100 mesh

Supplier: BeanTown Chemical

131395-100G 131395-25G 131395-5G
BT131395-100GEA 899.76 USD
BT131395-100G BT131395-25G BT131395-5G
Copper ≥99.999% (trace metals basis), powder -100 mesh
Copper

CAS: 7440-50-8; EC No: 231-159-6; MDL No: MFCD00010965; RTECS: GL5325000 UN No: UN3089; Haz Class: 4.1; Packing Group: III Powder; Molecular Formula: Cu; MW: 63.55 Melting Point: 1083.4°; Boiling Point: 2567° Density (g/mL): 8.94


  • RTECS: GL5325000
  • Powder

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/mL
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1 III
Merck Index: 13,02545

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR