Silver iodide ≥99.9%
Supplier: BeanTown Chemical
Synonyms:
Argentous iodide, Silver monoiodide
131380-100G
131380-25G
BT131380-100GEA
226.59
USD
BT131380-100G
BT131380-25G
Silver iodide ≥99.9%
Silver iodide
CAS: 7783-96-2; EC No: 232-038-0; MDL No: MFCD00003412; RTECS: VW4450000
UN No: UN3077; Haz Class: 9; Packing Group: III
Crystalline; Molecular Formula: AgI; MW: 234.77
Melting Point: 558°
Density (g/mL): 5.68
Light Sensitive
- RTECS: VW4450000
- Light Sensitive
- Crystalline
Formula:
AgI Melting Pt: 558 °C Density: 5.68 g/mL |
MDL Number:
MFCD00003412 CAS Number: 7783-96-2 EINECS: 232-038-0 ADR: 9,III |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...