Methylcyclohexane ≥99%

Supplier: BeanTown Chemical

131070-100G 131070-500G
BT131070-100GEA 39.97 USD
BT131070-100G BT131070-500G
Methylcyclohexane ≥99%
Formula: C₆H₁₁CH₃
Boiling Pt: 101 °C
Melting Pt: –126 °C
Density: 0.771 g/mL
Flash Pt: –4 °C (25 °F)
MDL Number: MFCD00001497
CAS Number: 108-87-2
EINECS: 203-624-3
UN: 2296
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f7f0d5a1c32138da16df4781897a9c0184e42b7&vid=39a9392c8629d0e2fc79134840e6d8a8dc8a76e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR