Lead(II) nitrate ≥99.0% ACS
Supplier: BeanTown Chemical
Synonyms:
Lead dinitrate
131000-2KG
131000-500G
BT131000-2KGEA
194.51
USD
BT131000-2KG
BT131000-500G
Lead(II) nitrate ≥99.0% ACS
Lead (II) nitrate
CAS: 10099-74-8; EC No: 233-245-9; MDL No: MFCD00011153; RTECS: OG2100000
UN No: UN1469; Haz Class: 5.1 (6.1); Packing Group: II
Crystalline; Linear Formula: Pb(NO3)2; MW: 331.20
Melting Point: 470° (decomposes)
Density (g/mL): 4.53
Hygroscopic
- RTECS: OG2100000
- Hygroscopic
- Crystalline
Formula:
Pb(NO₃)₂ MW: 331.21 g/mol Melting Pt: 470 °C (dec.) Density: 4.53 g/mL |
MDL Number:
MFCD00011153 CAS Number: 10099-74-8 EINECS: 233-245-9 UN: 1469 ADR: 5.1,II |
Specification Test Results
Assay | 99.0% min. |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...