Piperazine, anhydrous ≥99%
Supplier: BeanTown Chemical
130805-100G
130805-500G
BT130805-100GEA
34.46
USD
BT130805-100G
BT130805-500G
Piperazine, anhydrous ≥99%
Piperazine
CAS: 110-85-0; EC No: 203-808-3; MDL No: MFCD00005953; RTECS: TK7800000
UN No: UN2579; Haz Class: 8; Packing Group: III
Solid; Molecular Formula: C4H10O2; MW: 86.14
Melting Point: 109-112°; Boiling Point: 145-146°; Flash point: 88°C (190°F)
Density (g/mL): 1.11
Hygroscopic , Light Sensitive
- RTECS: TK7800000
- Hygroscopic, Light Sensitive
- Solid
Formula:
C₄H₁₀N₂ MW: 86.14 g/mol Boiling Pt: 145…146 °C Melting Pt: 109…112 °C Density: 1.11 g/mL Flash Pt: 88 °C (190 °F) |
MDL Number:
MFCD00005953 CAS Number: 110-85-0 EINECS: 203-808-3 UN: 2579 ADR: 8,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...