2-Furaldehyde ≥98.0% ACS

Supplier: BeanTown Chemical

Synonyms: 2-Furfurol, Furfural

130640-1KG 130640-250G
BT130640-1KGEA 103.24 USD
BT130640-1KG BT130640-250G
2-Furaldehyde ≥98.0% ACS
2-Furaldehyde

CAS: 98-01-1; EC No: 202-627-7; MDL No: MFCD00003229; RTECS: LT7000000 UN No: UN1199; Haz Class: 6.1 (3); Packing Group: II Liquid; Molecular Formula: C5H4O2; MW: 96.08 Melting Point: -36°; Boiling Point: 162°; Flash point: 60°C (140°F) Density (g/mL): 1.16; Refractive Index: 1.525 Air Sensitive


  • RTECS: LT7000000
  • Refractive index: 1.525
  • Air Sensitive
  • Liquid

Formula: C₅H₄O₂
MW: 96.09 g/mol
Boiling Pt: 162
Melting Pt: -39
Density: 1.16 g/mL
Flash Pt: 60 °C (140 °F)
MDL Number: MFCD00003229
CAS Number: 98-01-1
EINECS: 202-627-7
UN: 1199
ADR: 6.1 II
Merck Index: 13,04325

Order Now

Specification Test Results

Assay 98.0% min.
Residue ≤0.50%
Moisture ≤0.20%
Acidity ≤0.020eq/L


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR