Cyclopentane ≥95.0% for HPLC
Supplier: BeanTown Chemical
Synonyms:
Pentamethylene
130565-100ML
130565-2.5L
130565-500ML
BT130565-100MLEA
30.59
USD
BT130565-100ML
BT130565-2.5L
BT130565-500ML
Cyclopentane ≥95.0% for HPLC
Cyclopentane
CAS: 287-92-3; EC No: 206-016-6; MDL No: MFCD00001356; RTECS: GY2390000
UN No: UN1146; Haz Class: 3; Packing Group: II
Liquid; Molecular Formula: C5H10; MW: 70.14
Melting Point: -94°; Boiling Point: 50°; Flash point: -20°C (-4°F)
Density (g/mL): 0.751; Refractive Index: 1.405
- RTECS: GY2390000
- Refractive index: 1.405
- Liquid
Formula:
C₅H₁₀ MW: 70.13 g/mol Boiling Pt: 49 Melting Pt: -93.3 Density: 0.751 g/mL Flash Pt: –20 °C (–4 °F) |
MDL Number:
MFCD00001356 CAS Number: 287-92-3 EINECS: 206-016-6 UN: 1146 ADR: 3 II Merck Index: 13,02769 |
|
Specification Test Results
Assay | 95.0% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...