Ytterbium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.1 mm

Supplier: BeanTown Chemical

130125-25X25MM 130125-50X50MM
BT130125-25X25EA 136 USD
BT130125-25X25 BT130125-50X50
Ytterbium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.1 mm
Ytterbium

CAS: 7440-64-4; EC No: 231-173-2; MDL No: MFCD00011286 UN No: UN3178; Haz Class: 4.1; Packing Group: III Solid; Molecular Formula: Yb; MW: 173.04 Melting Point: 824°; Boiling Point: 1196° Density (g/mL): 6.54


  • Solid

Formula: Yb
Boiling Pt: 1196 °C
Melting Pt: 824 °C
Density: 6.54 g/mL
MDL Number: MFCD00011286
CAS Number: 7440-64-4
EINECS: 231-173-2
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR