Potassium cyanide ≥97%
Supplier: BeanTown Chemical
129825-100G
129825-500G
BT129825-100GEA
44.58
USD
BT129825-100G
BT129825-500G
Potassium cyanide ≥97%
Potassium cyanide
CAS: 151-50-8; EC No: 205-792-3; MDL No: MFCD00011397; RTECS: TS8750000
UN No: UN1680; Haz Class: 6.1; Packing Group: I
Solid; Linear Formula: KCN; MW: 65.12
Melting Point: 634°; Boiling Point: 1625°
Density (g/mL): 1.52
- RTECS: TS8750000
- Solid
Formula:
KCN MW: 65.12 g/mol Boiling Pt: 1625 °C Melting Pt: 634 °C Density: 1.52 g/mL |
MDL Number:
MFCD00011397 CAS Number: 151-50-8 EINECS: 205-792-3 UN: 1680 ADR: 6.1,I |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...