Vinyltrimethoxysilane ≥98%

Supplier: BeanTown Chemical

Synonyms: Trimethoxy(vinyl)silane

129420-100G 129420-500G
BT129420-100GEA 41.55 USD
BT129420-100G BT129420-500G
Vinyltrimethoxysilane ≥98%
Vinyltrimethoxysilane

CAS: 2768-02-7; EC No: 220-449-8; MDL No: MFCD00008605; RTECS: VV6770000 UN No: UN1993; Haz Class: 3; Packing Group: III Liquid; Linear Formula: H2C=CHSi(OCH3)3; Molecular Formula: C5H12O3Si; MW: 148.24 Melting Point: -97°; Boiling Point: 122-124°/76 mmHg; Flash point: 22°C (72°F) Density (g/mL): 0.968; Refractive Index: 1.392 Moisture Sensitive


  • RTECS: VV6770000
  • Refractive index: 1.392
  • Moisture Sensitive
  • Liquid

Formula: C₅H₁₂O₃Si
MW: 148.23 g/mol
Boiling Pt: 122…124 °C/76 mmHg
Melting Pt: –97 °C
Density: 0.968 g/mL
Flash Pt: 22 °C (72 °F)
MDL Number: MFCD00008605
CAS Number: 2768-02-7
EINECS: 220-449-8
ADR: 3,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR