Methanol-[D₄] ≥100% (Isotopic)
Supplier: BeanTown Chemical
Synonyms:
Methanol-[²H₄], Hydroxy methane-D4, Methanol-d4, Methyl-D3 alcohol-D, Carbinol-D4, Wood alcohol-D4, Methyl alcohol-D4, Tetradeuteromethanol
129405-10EACH
129405-2EACH
BT129405-10EAEA
476
USD
BT129405-10EA
BT129405-2EACH
Methanol-[D₄] ≥100% (Isotopic)
Methanol-[D4]
CAS: 811-98-3; EC No: 212-378-6; MDL No: MFCD00044637
UN No: UN1230; Haz Class: 3; Packing Group: II
Liquid, in prescored ampules; Linear Formula: CD3OD; MW: 36.07
Melting Point: -98°; Boiling Point: 65.4°; Flash point: 11°C (54°F)
Density (g/mL): 0.888; Refractive Index: 1.326
Hygroscopic
- Refractive index: 1.326
- Hygroscopic
- Liquid, in prescored ampules
Formula:
D₃COD MW: 36.01 g/mol Boiling Pt: 65.3 Melting Pt: -99 Density: 0.888 g/mL Flash Pt: 11 °C (54 °F) |
MDL Number:
MFCD00044637 CAS Number: 811-98-3 EINECS: 212-378-6 UN: 1230 ADR: 3 II |
Specification Test Results
Assay | 100% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...