4,4′-Azo-bis(4-cyanovaleric acid) ≥98%
Supplier: BeanTown Chemical
129380-100G
129380-25G
BT129380-100GEA
196.18
USD
BT129380-100G
BT129380-25G
4,4′-Azo-bis(4-cyanovaleric acid) ≥98%
4,4′-Azo-bis(4-cyanovaleric acid)
CAS: 2638-94-0; EC No: 220-135-0; MDL No: MFCD00002799; RTECS: YV6190500
UN No: UN1325; Haz Class: 4.1; Packing Group: II
Solid; Molecular Formula: C12H16N4O4 ; MW: 280.28
Melting Point: 118-125° (decomposes)
Light Sensitive
- RTECS: YV6190500
- Light Sensitive
- Solid
Formula:
C₁₂H₁₆N₄O₄ MW: 280.284 g/mol Melting Pt: 118…125 °C (dec.) |
MDL Number:
MFCD00002799 CAS Number: 2638-94-0 EINECS: 220-135-0 ADR: 4.1 II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...