Potassium chromate ≥99.0% ACS
Supplier: BeanTown Chemical
128065-100G
128065-500G
BT128065-100GEA
46.2
USD
BT128065-100G
BT128065-500G
Potassium chromate ≥99.0% ACS
Potassium chromate
CAS: 7789-00-6; EC No: 232-140-5; MDL No: MFCD00011368; RTECS: GB2940000
UN No: UN3087; Haz Class: 5.1 (6.1); Packing Group: III
Crystalline; Linear Formula: K2CrO4; MW: 194.19
Melting Point: 971°
Density (g/mL): 2.73
- RTECS: GB2940000
- Crystalline
Formula:
K₂CrO₄ MW: 194.19 g/mol Melting Pt: 971 °C Density: 2.73 g/mL |
MDL Number:
MFCD00011368 CAS Number: 7789-00-6 EINECS: 232-140-5 UN: 3087 ADR: 5.1,III |
Specification Test Results
Assay | 99.0% min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...