Nickel(II) oxalate dihydrate

Supplier: BeanTown Chemical

Synonyms: Nickel oxalate dihydrate

128060-250G 128060-50G
BT128060-250GEA 234 USD
BT128060-250G BT128060-50G
Nickel(II) oxalate dihydrate
Nickel(II) oxalate dihydrate

CAS: 6018-94-6; EC No: 208-933-7; MDL No: MFCD00167155 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Linear Formula: NiC2O4·2H2O; MW: 182.74


  • Powder

Formula: C₂H₄NiO₆
MDL Number: MFCD00167155
CAS Number: 6018-94-6
EINECS: 208-933-7
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR