Quinidine

Supplier: BeanTown Chemical

Synonyms: (+)-Quinidine

127895-1G 127895-25G 127895-5G
BT127895-1GEA 25 USD
BT127895-1G BT127895-25G BT127895-5G
Quinidine
Quinidine

CAS: 56-54-2; EC No: 200-279-0; MDL No: MFCD00135581; RTECS: VA4725000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Crystalline; Molecular Formula: C20H24N2O2; MW: 324.42 Melting Point: 168-172° Light Sensitive


  • RTECS: VA4725000
  • Light Sensitive
  • Crystalline

Formula: C20H24N2O2
MW: 324.42 g/mol
Melting Pt: 168…172 °C
MDL Number: MFCD00135581
CAS Number: 56-54-2
EINECS: 200-279-0
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR