Nickel diiodide, anhydrous ≥99.5% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Nickel iodide (NiI2)
, Nickel (II) iodide
127465-5G
127465-25G
BT127465-5GEA
48.16
USD
BT127465-5G
BT127465-25G
Nickel diiodide, anhydrous ≥99.5% (trace metals basis)
Nickel diiodide
- Hygroscopic, Light Sensitive
- -8 Mesh Powder
Formula:
I₂Ni MW: 312.5 g/mol Melting Pt: 797 °C Density: 5.834 g/mL |
MDL Number:
MFCD00016264 CAS Number: 13462-90-3 EINECS: 236-666-6 UN: 3077 ADR: 8,III |
Specification Test Results
Assay | 99.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...