Gadolinium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.025 mm
Supplier: BeanTown Chemical
126965-25X25MM
126965-50X50MM
BT126965-25X25EA
120
USD
BT126965-25X25
BT126965-50X50
Gadolinium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.025 mm
Gadolinium
CAS: 7440-54-2; EC No: 231-162-2; MDL No: MFCD00011022; RTECS: LW3850000
UN No: UN3178; Haz Class: 4.1; Packing Group: II
Solid; Molecular Formula: Gd; MW: 157.25
Melting Point: 1313°; Boiling Point: 3273°
Density (g/mL): 7.886
- RTECS: LW3850000
- Solid
Formula:
Gd Boiling Pt: 3273 °C Melting Pt: 1313 °C Density: 7.886 g/mL |
MDL Number:
MFCD00011022 CAS Number: 7440-54-2 EINECS: 231-162-2 ADR: 4.1,II |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...