Acetic anhydride ≥99%
Supplier: BeanTown Chemical
Synonyms:
Acetyl oxide, Acetyl acetate
125580-1KG
125580-2.5KG
BT125580-1KGEA
67.47
USD
BT125580-1KG
BT125580-2.5KG
BT125580-250G
Acetic anhydride ≥99%
Acetic anhydride
CAS: 108-24-7; EC No: 203-564-8; MDL No: MFCD00008705; RTECS: AK1925000
UN No: UN1715; Haz Class: 8 (3); Packing Group: II
Liquid; Linear Formula: (CH3CO)2O; MW: 102.09
Melting Point: -73°; Boiling Point: 138-140°; Flash point: 49°C (120°F)
Density (g/mL): 1.08; Refractive Index: 1.390
Moisture Sensitive
- RTECS: AK1925000
- Refractive index: 1.390
- Moisture Sensitive
- Liquid
Formula:
(CH₃CO)₂O MW: 102.09 g/mol Boiling Pt: 138…140 °C Melting Pt: –73 °C Density: 1.08 g/mL Flash Pt: 49 °C (120 °F) |
MDL Number:
MFCD00008705 CAS Number: 108-24-7 EINECS: 203-564-8 UN: 1715 ADR: 8,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...