Gallium antimonide ≥99.99%
Supplier: BeanTown Chemical
125065-1G
125065-5G
BT125065-1GEA
53
USD
BT125065-1G
BT125065-5G
Gallium antimonide ≥99.99%
Gallium antimonide
CAS: 12064-03-8; EC No: 235-058-8; MDL No: MFCD00016101
UN No: UN1549; Haz Class: 6.1; Packing Group: III
6mm & Down Pieces; Molecular Formula: GaSb; MW: 191.47
Melting Point: 710°
Density (g/mL): 5.619
- 6mm & Down Pieces
Formula:
GaSb Melting Pt: 710 °C Density: 5.619 g/mL |
MDL Number:
MFCD00016101 CAS Number: 12064-03-8 EINECS: 235-058-8 UN: 1549 ADR: 6.1,III |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...