Lead(II) carbonate ≥98% ACS

Supplier: BeanTown Chemical

Synonyms: Lead carbonat

124675-100G 124675-500G
BT124675-100GEA 57.95 USD
BT124675-100G BT124675-500G
Lead(II) carbonate ≥98% ACS
Lead (II) carbonate

CAS: 598-63-0; EC No: 209-943-4; MDL No: MFCD00064631 UN No: UN2291; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: PbCO3 ; MW: 267.20


  • Powder

Formula: PbCO₃
MW: 267.21 g/mol
MDL Number: MFCD00064631
CAS Number: 598-63-0
EINECS: 209-943-4
UN: 2291
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%
Insoluble in dilute CH3COOH ≤0.02%
Chloride (Cl) ≤0.002%
Nitrate & nitrite Passes test
Cadmium (Cd) ≤0.002%
Calcium (Ca) ≤0.01%
Iron (Fe) ≤0.005%
Potassium (K) ≤0.02%
Sodium (Na) ≤0.05%
Zinc (Zn) ≤0.003%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR