1-Butanesulfonyl chloride ≥98%

Supplier: BeanTown Chemical

Synonyms: 1-Butanesulphonylchloride, 1-Butanesulfonylchloride

124115-100G 124115-25G 124115-5G
BT124115-100GEA 253 USD
BT124115-100G BT124115-25G BT124115-5G
1-Butanesulfonyl chloride ≥98%
1-Butanesulphonyl chloride

CAS: 2386-60-9; EC No: 219-204-8; MDL No: MFCD00007464; RTECS: EK5950000 UN No: UN3265; Haz Class: 8; Packing Group: II Liquid; Linear Formula: CH3(CH2)3SO2Cl; Molecular Formula: C4H9ClO2S; MW: 156.63 Boiling Point: 80-81°/9 mmHg; Flash point: 79°C (174°F) Density (g/mL): 1.208; Refractive Index: 1.454 Moisture Sensitive


  • RTECS: EK5950000
  • Refractive index: 1.454
  • Moisture Sensitive
  • Liquid

Formula: C₄H₉ClO₂S
MW: 156.63 g/mol
Boiling Pt: 208
Melting Pt: -25
Density: 1.208 g/mL
Flash Pt: 79 °C (174 °F)
MDL Number: MFCD00007464
CAS Number: 2386-60-9
EINECS: 219-204-8
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR