Acetone ≥99.5% semiconductor grade
Supplier: BeanTown Chemical
CAS: 67-64-1; EC No: 200-662-2; MDL No: MFCD00008765; RTECS: AL3150000
UN No: UN1090; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3COCH3; MW: 58.08
Melting Point: -94°; Boiling Point: 56°; Flash point: -17°C (1°F)
Density (g/mL): 0.791; Refractive Index: 1.359
- RTECS: AL3150000
- Refractive index: 1.359
- Liquid
Formula:
CH₃COCH₃ Boiling Pt: 56 °C Melting Pt: –94 °C Density: 0.791 g/mL Flash Pt: –17 °C (1 °F) |
MDL Number:
MFCD00008765 CAS Number: 67-64-1 EINECS: 200-662-2 UN: 1090 ADR: 3,II |
|
Specification Test Results
Assay | 99.5% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...