Iodoacetamide ≥98%

Supplier: BeanTown Chemical

122600-100G 122600-25G 122600-5G
BT122600-100GEA 265.08 USD
BT122600-100G BT122600-25G BT122600-5G
Iodoacetamide ≥98%
Iodoacetamide

CAS: 144-48-9; EC No: 205-630-1; MDL No: MFCD00008028; RTECS: AC4200000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Crystalline; Linear Formula: ICH2CONH2; Molecular Formula: C2H4IO; MW: 184.96 Melting Point: 92-95° Light Sensitive


  • RTECS: AC4200000
  • Light Sensitive
  • Crystalline

Formula: ICH₂CONH₂
Melting Pt: 92…95 °C
MDL Number: MFCD00008028
CAS Number: 144-48-9
EINECS: 205-630-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR