Chlorotitanium triisopropoxide ≥95%
Supplier: BeanTown Chemical
122510-100G
122510-25G
122510-500G
BT122510-100GEA
142.87
USD
BT122510-100G
BT122510-25G
BT122510-500G
Chlorotitanium triisopropoxide ≥95%
Chlorotitanium triisopropoxide
CAS: 20717-86-6; MDL No: MFCD00009861
UN No: UN2921; Haz Class: 8 (4.1); Packing Group: II
Solid; Linear Formula: [(CH3)2CHO]3TiCl; Molecular Formula: C9H21ClO3Ti; MW: 260.58
Melting Point: 35-40°; Flash point: 22°C (72°F)
Density (g/mL): 1.091
Moisture Sensitive
- Moisture Sensitive
- Solid
Formula:
C₉H₂₁ClO₃Ti Melting Pt: 35…40 °C Density: 1.091 g/mL Flash Pt: 22 °C (72 °F) |
MDL Number:
MFCD00009861 CAS Number: 20717-86-6 UN: 2921 ADR: 8,II |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...