Benzene ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: Cyclohexatriene

121915-1L 121915-4L 121915-500ML
BT121915-1LEA 66.2 USD
BT121915-1L BT121915-4L BT121915-500ML
Benzene ≥99.0% ACS
Benzene

CAS: 71-43-2; EC No: 200-753-7; MDL No: MFCD00003009; RTECS: CY1400000 UN No: UN1114; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C6H6; MW: 78.11 Melting Point: 5.5°; Boiling Point: 80°; Flash point: -11°C (12°F) Density (g/mL): 0.874; Refractive Index: 1.501


  • RTECS: CY1400000
  • Refractive index: 1.501
  • Liquid

Formula: C₆H₆
MW: 78.11 g/mol
Boiling Pt: 80 °C
Melting Pt: 5.5 °C
Density: 0.874 g/mL
Flash Pt: –11 °C (12 °F)
MDL Number: MFCD00003009
CAS Number: 71-43-2
EINECS: 200-753-7
UN: 1114
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2c0146712ab33142aeeb3843b02381e6bda4028&vid=cae270bfe08ae6ab4b14d9e1775b89a82cfeec1f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR